Alfa Aesar™ Molybdenum(VI) oxide bis(2,4-pentanedionate), 99%

Manufacturer: Alfa Aesar™ 4414018

Quantity 50g

 View more versions of this product

Catalog No. AA4414018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17524-05-9
    Molecular Formula C10H14MoO6
    Formula Weight 326.16
    MDL Number MFCD00011506
    Synonym Bis(acetylacetonato)dioxomolybdenum(VI); Molybdenyl acetylacetonate
    Chemical Name or Material Molybdenum (VI) oxide bis (2,4-pentanedionate)

    Specifications

    Formula Weight 326.16
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Air Sensitive
    Color Pale yellow
    Molecular Formula C10H14MoO6
    MDL Number MFCD00011506
    Melting Point ≈185°C (dec.)
    Health Hazard 1 Warning
    Physical Form Powder
    CAS 17524-05-9
    Synonym Bis(acetylacetonato)dioxomolybdenum(VI); Molybdenyl acetylacetonate
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Molybdenum (VI) oxide bis (2,4-pentanedionate)
    Assay 99%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit