Alfa Aesar™ Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max

Manufacturer: Alfa Aesar™ 1081222

Quantity 100g

 View more versions of this product

Catalog No. AA1081222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1313-27-5
    Formula Weight 143.94
    MDL Number MFCD00003469
    UN Number UN3288
    Merck Index 14,6239
    Chemical Name or Material Molybdenum(VI) oxide
    Grade Puratronic™

    Specifications

    Formula Weight 143.94
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H300-H351-H319-H335
    Fatal if swallowed.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Odor Odorless
    UN Number UN3288
    Merck Index 14,6239
    Boiling Point 1155°C (subl.)
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Physical Form Powder
    MDL Number MFCD00003469
    CAS 1313-27-5
    Quantity 100g
    Density 4.692g/mL
    Color White to gray
    Melting Point 795°C
    RTECS Number QA4725000
    Chemical Name or Material Molybdenum(VI) oxide
    Assay 99.998%, (metals basis excluding W), W 300ppm max.
    Grade Puratronic™

    GHS H Statement
    H300-H351-H319-H335
    Fatal if swallowed.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit