Alfa Aesar™ Molybdenum(VI) sulfide dihydrate

Manufacturer: Alfa Aesar™ 3274106

Quantity 5g

 View more versions of this product

Catalog No. AA3274106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12033-29-3
    Formula Weight 228.13 (192.13 Anhydrous)
    MDL Number MFCD00014226
    Chemical Name or Material Molybdenum(VI) sulfide dihydrate

    Specifications

    Formula Weight 228.13 (192.13 Anhydrous)
    TSCA TSCA
    Odor Odorless
    Physical Form Powder
    CAS 12033-29-3
    MDL Number MFCD00014226
    Quantity 5g
    Chemical Name or Material Molybdenum(VI) sulfide dihydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit