Alfa Aesar™ Molybdenum wire, 0.013mm (0.00052 in.) dia., hard, 99.8% (metals basis)

Microelectronics, high temp

Manufacturer: Alfa Aesar™ 40669G1

Quantity 1m

 View more versions of this product

Catalog No. AA40669G1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-98-7
    MDL Number MFCD00003465
    Chemical Name or Material Molybdenum wire, 0.013mm (0.00052 in.) dia., Hard

    Specifications

    Physical Form Wire
    CAS 7439-98-7
    Quantity 1m
    TSCA TSCA
    MDL Number MFCD00003465
    Weight ≈0.0014g/m
    Chemical Name or Material Molybdenum wire, 0.013mm (0.00052 in.) dia., Hard
    Assay 99.8%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit