Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Molybdenum wire, 0.07mm (0.003 in.) dia., 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 00876H4

Quantity 100m

 View more versions of this product

Catalog No. AA00876H4

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7439-98-7
MFCD00003465
Molybdenum wire, 0.07mm (0.003 in.) dia.

Specifications

≈0.04g/m
7439-98-7
MFCD00003465
Molybdenum wire, 0.07mm (0.003 in.) dia.
100m
Wire
Air Sensitive
99.95%, (metals basis)

Safety and Handling

RTECSNumber : QA4680000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit