Alfa Aesar™ Molybdenum wire, 0.127mm (0.005 in.) dia., annealed, 99.95% (metals basis)

Manufacturer: Alfa Aesar™ 10037I1

Quantity 1000m

 View more versions of this product

Catalog No. AA10037I1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-98-7
    MDL Number MFCD00003465
    Chemical Name or Material Molybdenum wire, 0.127mm (0.005 in.) dia., Annealed, Temper: soft

    Specifications

    CAS 7439-98-7
    Weight ≈0.13g/m
    TSCA TSCA
    Physical Form Wire
    MDL Number MFCD00003465
    Quantity 1000m
    Chemical Name or Material Molybdenum wire, 0.127mm (0.005 in.) dia., Annealed, Temper: soft
    Assay 99.95%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit