Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum wire, 99.94% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
100m
25m
5m
7439-98-7
Mo
95.95
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2
23932
CHEBI:28685
molybdenum
[Mo]
7439-98-7
Mo
95.95
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum, elemental,6+ ion,mchvl,vi ion,molybdenum, metallic,metco 63,atom,molybdenum, ion 6+,tsm1,amperit 106.2
23932
CHEBI:28685
molybdenum
[Mo]

Form: approx. 8.0g/m, Temper: soft

Quantity 25m
Appearance Dark gray wire
Assay Percent Range 99.94% (Metals basis)
Odor Odorless
Physical Form Wire, ≈8.0g/m,Temper: soft
Color Gray
Chemical Name or Material Molybdenum

EINECSNumber : 231-107-2

TSCA : Yes

Recommended Storage : Ambient temperatures