Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenyl acetylacetonate, 97%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
17524-05-9
C10H14MoO6
326.15
2, 4-Pentanedione, metal derivative
17524-05-9
C10H14MoO6
326.15
2, 4-Pentanedione, metal derivative
Oxidation reagent
Infrared Spectrum Authentic
Elemental Analysis 96.0% min.
Melting Point 184.0°C
Packaging Glass bottle
Quantity 250g
Color Undesignated
Assay Percent Range 97%
Solubility Information Solubility in water: 85g/L (20°C). Other solubilities: 0.31 g/100g alcohol (20°C), 1.84 g/100g benzene (20°C)
Formula Weight 326.15
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Molybdenyl acetylacetonate
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plen

GHS Signal Word: Warning

EINECSNumber : 241-522-