Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdic Acid, 85% (Powder/Certified ACS), Fisher Chemical

Click to view available options
Quantity:
500g
Packaging:
Glass Bottle
12054-85-2
(NH4)6MO7O24 · 4 H2O
MFCD00036287
Ammonium Molybdate
12054-85-2
(NH4)6MO7O24 · 4 H2O
MFCD00036287
Ammonium Molybdate

This reagent consists largely of ammonium molybdate

Packaging Glass Bottle
Quantity 500g
Melting Point 190°C
Density 2.49g/cm³
Identification Pass Test
Color Yellow
Assay Percent Range ≥85.0 %
Solubility Information 0.01 max. Insoluble in NH4OH
Physical Form Powder/Solid
Percent Purity ≥85.0%
Grade Certified ACS
Chemical Name or Material Molybdic Acid, 85%
Show More Show Less