Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Molybdic acid, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Color | White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 100 g |
Merck Index | 15,6326 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in nh4oh, h2so4 and fixed alkalies |
Formula Weight | 161.96 |
Percent Purity | ≥85.0% (MoO3) |
Grade | ACS Reagent |
Packaging | Plastic bottle |
Specific Gravity | 3.1 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes serious eye irritation. May cause damage to organs through prolonged or repeated exposure. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Wear eye protection/face protection. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/ |
EINECSNumber | 231-970-5 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction