Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdic acid, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g,100g
500g,500g
Packaging:
Plastic bottle,Plastic bottle
7782-91-4
H2MoO4
161.96
MFCD00010882
VLAPMBHFAWRUQP-UHFFFAOYSA-L
molybdic acid,molybdic vi acid,unii-i96n991j1n,dihydroxy dioxo molybdenum,molybdic acid acs,dihydroxy-dioxomolybdenum,molybdate moo42-, dihydrogen, t-4,dihydroxidodioxidomolybdenum,aronis24112,moo2 oh 2
82208
CHEBI:25371
dihydroxy(dioxo)molybdenum
O[Mo](=O)(=O)O
7782-91-4
H2MoO4
161.96
MFCD00010882
VLAPMBHFAWRUQP-UHFFFAOYSA-L
molybdic acid,molybdic vi acid,unii-i96n991j1n,dihydroxy dioxo molybdenum,molybdic acid acs,dihydroxy-dioxomolybdenum,molybdate moo42-, dihydrogen, t-4,dihydroxidodioxidomolybdenum,aronis24112,moo2 oh 2
82208
CHEBI:25371
dihydroxy(dioxo)molybdenum
O[Mo](=O)(=O)O
Oxidation reagent
Density 3.1000g/mL,3.1000g/mL
Specific Gravity 3.1,3.1
Quantity 500g,500g
Packaging Plastic bottle,Plastic bottle
Color White to Yellow,White through Yellow
Additional Information contains ammonium molybdate
Merck Index 15, 6326
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in nh4oh,h2so4 and fixed alkalies
Formula Weight 161.96
Physical Form Powder
Percent Purity ≥85% (MoO3)
Grade Analysis
Chloride 100ppm max
Chlorine (Cl) 100ppm max.
Heavy Metals (as Pb) 30ppm max.
Iron (Fe) 50ppm max.
Phosphate 5ppm max.
Sulfate 2000ppm max.
Chemical Name or Material Molybdic acid, p.a.
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.

GHS P Statement
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/

GHS Signal Word: Warning

EINECSNumber : 231-970-5

TSCA : TSCA