Learn More
MON1A Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 237721AP
Catalog No. 50-173-4120
Add to cart
Description
Description
MON1A is a cytosolic protein that peripherally associates with the ER. MON1A plays an important role in membrane trafficking through the secretory apparatus. MON1A affects different steps in the secretory pathway including endoplasmic reticulum-to-Golgi traffic. MON1A encodes several isoforms with MW about 62, 55, 45 and 34 kDa.
Specifications
Specifications
MON1A
0.59 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86VX9
MON1 homolog A (yeast), MON1A, SAND1
Rabbit
IgG
150 μL
-20°C
Primary
84315
Western Blot
Unconjugated
MON1A
Liquid
MON1A
MON1A Fusion Protein Ag6900
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human