Alfa Aesar™ Monel™ 400 wire, 0.25mm (0.01 in.) dia.

Manufacturer: Alfa Aesar™ 45065TB

Quantity 113g

 View more versions of this product

Catalog No. AA45065TB

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD09910536
    Chemical Name or Material Monel™ 400 wire, 0.25mm (0.01 in.) dia.

    Specifications

    Quantity 113g
    TSCA TSCA
    Physical Form Wire
    MDL Number MFCD09910536
    Weight ≈0.4339g/m
    Chemical Name or Material Monel™ 400 wire, 0.25mm (0.01 in.) dia.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit