Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monensin sodium salt, 50mg/mL in methanol, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
5 x 1mL
22373-78-0
C36H61NaO11
692.863
MFCD00077826
XOIQMTLWECTKJL-JXDONITHSA-M
133612076
sodium;(3S,4R)-4-[(2R,5S,7R,8S,9R)-2-[(2S,5R)-5-ethyl-5-[(2S,3R,5S)-5-[(2R,3R,5S,6S)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]oxolan-2-yl]-7-hydroxy-2,8-dimethyl-1,10-dioxaspiro[4.5]decan-9-yl]-3-methoxy-2-methylpentanoate
CCC1(CCC(O1)C2(CCC3(O2)CC(C(C(O3)C(C)C(C(C)C(=O)[O-])OC)C)O)C)C4C(CC(O4)C5C(CC(C(O5)(CO)O)C)C)C.[Na+]
22373-78-0
C36H61NaO11
692.863
MFCD00077826
XOIQMTLWECTKJL-JXDONITHSA-M
133612076
sodium;(3S,4R)-4-[(2R,5S,7R,8S,9R)-2-[(2S,5R)-5-ethyl-5-[(2S,3R,5S)-5-[(2R,3R,5S,6S)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]oxolan-2-yl]-7-hydroxy-2,8-dimethyl-1,10-dioxaspiro[4.5]decan-9-yl]-3-methoxy-2-methylpentanoate
CCC1(CCC(O1)C2(CCC3(O2)CC(C(C(O3)C(C)C(C(C)C(=O)[O-])OC)C)O)C)C4C(CC(O4)C5C(CC(C(O5)(CO)O)C)C)C.[Na+]
UN Number UN1230
Physical Form Liquid
Concentration 50mg/mL in methanol
Quantity 5 x 1mL
Chemical Name or Material Monensin sodium salt, sterile-filtered

GHS H Statement
H225-H301-H311-H331-H370
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL

RTECSNumber : JH2830000

storageNote1 : −30° to −10°C