Learn More
Novus Biologicals™ Monkey ACTH ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP266414
Description
The Monkey ACTH ELISA Kit (Colorimetric) measures monkey ACTH in serum, plasma and other biological fluids.Specifications
15.63 - 1000 pg/mL | |
This kit recognizes Monkey ACTH in samples. No significant cross-reactivity or interference between Monkey ACTH and analogues was observed. | |
5443 | |
Monkey | |
1 Kit | |
Storage is content dependent. |
9.38 pg/mL | |
ELISA | |
POMC | |
ELISA Kit (Colorimetric) | |
Biological Fluids,Plasma,Serum | |
ACTH |