Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Monkey C-Peptide ELISA Kit (Colorimetric)
AD7C-NTP Colorimetric ELISA Kit for quantitative evaluation of C-peptide
Supplier: Novus Biologicals™ NBP2599571KIT
Description
The Monkey C-Peptide ELISA Kit (Colorimetric) measures monkey C-Peptide in serum, plasma, tissue homogenates.Specifications
0.7 to 10ng/mL | |
ELISA Kit (Colorimetric) | |
Detection of cynomologus monkey C-Peptide | |
ELISA | |
INS | |
1 Kit | |
Lipid and Metabolism | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
C-Peptide |
0.3ng/mL | |
Plasma, Serum, Tissue Homogenates | |
Monkey | |
3630 | |
Assay plate, Standard, HRP-conjugate, Wash Buffer (20X concentrate), Substrate A, Substrate B, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual | |
RUO | |
50 to 100 μL | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction