Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Monkey C-Peptide ELISA Kit (Colorimetric)

AD7C-NTP Colorimetric ELISA Kit for quantitative evaluation of C-peptide

$743.50

Specifications

Species Monkey
Sensitivity 0.3ng/mL
Sample Type Serum, plasma, tissue homogenates.
Research Discipline Lipid and Metabolism
Synonym IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB599571KT View Documents Novus Biologicals™
NBP2599571KIT
1 Kit Each for $743.50
Add to cart
 
Description

Description

The Monkey C-Peptide ELISA Kit (Colorimetric) measures monkey C-Peptide in serum, plasma, tissue homogenates.
Specifications

Specifications

Monkey
Serum, plasma, tissue homogenates.
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
C-Peptide
3630
ELISA Kit (Colorimetric)
50 to 100μL
0.3ng/mL
Lipid and Metabolism
Store the unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
ELISA
INS
0.7 to 10ng/mL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit