Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Monkey Diet, Semi-purified, Pellet

Semi-purified monkey diet. Contains whole wheat, corn meal, casein and other nutritious items.

Manufacturer:  MP Biomedicals 0296033520

 View more versions of this product

Catalog No. MP296033520


Add to cart

Description

Description

  • Semi-purified monkey diet
  • Contains whole wheat, corn meal, casein, MP vitamin mix and mineral mix
Specifications

Specifications

Yes
0.3048 m
Animal Diet
Pellet
Room Temperature
1 ft.
20 kg
Autoclavable
Non-sterile
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.