Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Monkey Insulin ELISA Kit (Colorimetric)
CMV antibody (IgM) Colorimetric ELISA Kit useful for study of Infection
Supplier: Novus Biologicals™ NBP2600761KIT
Description
The Monkey Insulin ELISA Kit (Colorimetric) measures monkey Insulin in serum, plasma.Specifications
8 to 100uIU/mL | |
ELISA Kit (Colorimetric) | |
Detection of monkey INS | |
ELISA | |
INS | |
RUO | |
50 to 100 μL | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin |
5uIU/mL | |
Plasma, Serum | |
Monkey | |
3630 | |
1 Kit | |
Lipid and Metabolism | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
Insulin |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction