Learn More
Novus Biologicals™ Monkey Lipocalin-2/NGAL ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP262164
Catalog No. NBP262164
Add to Cart
Description
Description
The Monkey Lipocalin-2/NGAL ELISA Kit (Colorimetric) measures monkey Lipocalin-2/NGAL in monkey urine, plasma, serum, tissue extracts, culture supernatants.
Specifications
Specifications
10-200 pg/ml
ELISA Kit (Colorimetric)
ELISA
LCN2
1 Kit
Lipocalin-2/NGAL
1.5 pg/ml
Cell Culture Supernatant, Monkey Urine, Plasma, Serum, Tissue Extracts
3934
Monkey
Store at 4°C
Product Suggestions
Product Suggestions