Learn More
Novus Biologicals™ Monkey Osteoprotegerin/TNFRSF11B ELISA Kit (Colorimetric)
Monkey Osteoprotegerin/TNFRSF11B ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276686
Catalog No. NBP276686
Add to cart
Description
Description
The Monkey Osteoprotegerin/TNFRSF11B ELISA Kit (Colorimetric) measures monkey Osteoprotegerin/TNFRSF11B in serum, plasma and other biological fluids.
Specifications
Specifications
0.156 to 10 ng/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
TNFRSF11B
Biological Fluids, Plasma, Serum
4982
0.094 ng/mL
Monkey
1 Kit