Learn More
BioworldSupplier Diversity Partner Monkey Serum, 20 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306110871
Gemini monkey serum is aseptically collected from Macaques (Rhesus and/or Cynomolgus) that have been tested for Herpes B virus and found to be non-reactive. Herpes B virus is common and usually harmless to Macaques, but can be fatal in humans. This product should be handled with extreme care.Monkey serum can be infectious despite safety testing.SpecificationsAseptically collectedShips on dry ice.
Catalog No. 50-254-200