Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Monkey total TNF-alpha Platinum ELISA Kit Non-distribution product as customer accommodation.

For the quantitative detection of monkey TNF-alpha.

Manufacturer:  Invitrogen™ BMS654TEN

 View more versions of this product

Catalog No. 50-112-5478


Add to cart

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey TNF-alpha
Biotin-Conjugate anti-monkey TNF-alpha polyclonal antibody
Streptavidin-HRP
Monkey TNF-alpha Standard lyophilized, 3600 U/ml upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Tumor Necrosis Factor is a polypeptide cytokine produced by monocytes and macrophages. It functions as a multipotent modulator of immune response and further acts as a potent pyrogen. TNF-alpha circulates throughout the body responding to stimuli (infectious agents or tissue injury), activating neutrophils, altering the properties of vascular endothelial cells, regulating metabolic activities of other tissues, as well as exhibiting tumoricidal activity by inducing localized blood clotting. TNF-alpha also inhibits lipoprotein lipase activity resulting in cachexia, a physical wasting condition. Activation of B-cells by the Epstein Barr virus can be inhibited by TNF-alpha. Due to its varied actions throughout the immune system, TNF-alpha may play a role in the pathogenesis of many disease states. TNF-alpha production is mediated by the action of lymphokines and endotoxins on the macrophage. Purified monocytes produce TNF-alpha within four hours of stimulation by recombinant IL-2 and there is some in vitro evidence to suggest that TNF-alpha is expressed at high levels and with prolonged kinetics in T cells stimulated by both CD2 and CD28. Secretion of TNF-alpha is enhanced by gamma interferon. TNF then induces or enhances the specific production of Class I MHC antigen, GM-CSF, and IL-1.
Specifications

Specifications

P59695,P79337
Biotin
100126739, 102139631
Adhesive Plate Covers
10 x 96 tests
6.0 U/mL
50μL
Monkey
1 hr 20 min
Baboon, Cynomolgus Monkey
Cachectin, Tumor necrosis factor alpha, DIF
8%
28.1-1,800 U/mL
ELISA
Colorimetric Microplate Reader
ELISA Kit
RUO
Serum,Supernatant
Serum,50 μL|Supernatant,50 μL
2-8°C
3 hr 10 min
TNF
8%
HRP
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only