Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monkeypox Virus B21R Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326105G
Description
Monkeypox Virus B21R Polyclonal antibody specifically detects Monkeypox Virus B21R in Virus samples. It is validated for Western BlotSpecifications
Monkeypox Virus B21R | |
Polyclonal | |
Western Blot | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Virus | |
Purified |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Monkeypox Virus B21R | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction