Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mono-6-O-(p-toluenesulfonyl)-beta-cyclodextrin 85.0+%, TCI America™
Click to view available options
Quantity:
200mg
Specifications
Color | White |
Quantity | 200mg |
Formula Weight | 1289.17 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥85.0% (HPLC) |
Chemical Name or Material | Mono-6-O-(p-toluenesulfonyl)-beta-cyclodextrin |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator