Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mono-Methyl Lysine Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116817
Description
Positive Samples: HeLa, MCF7, NIH/3T3, C6, Mouse liver, Rat liver.
Methylated lysine is derivative of the amino acid residue lysine where the sidechain ammonium group has been methylated one or more times.
Specifications
Mono-Methyl Lysine | |
Polyclonal | |
Unconjugated | |
monomethyl lysine | |
A synthetic peptide corresponding to a sequence containing monomethylated K. | |
100 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, Western Blot | |
1.01 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Chemical, Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction