Learn More
GE Healthcare Mono Q HR 16/10 Available on GSA/VA Contract for Federal Government customers only.
Offers high capacity during the polishing step in purification of biomolecules. GE Healthcare Mono Q HR 16/10 is a strong anion exchanger packed with MonoBeads in high-volume beds. It is an excellent choice when high purity is needed.
Manufacturer:  GE Healthcare 17050601
Catalog No. 45-002-363
Add to cart
Description
Description
Mono Q HR 16/10 is a pre-packed column with a strong anion exchanger and MonoBeads in a 20ml bed for high capacity. It is ideal for the polishing step in purification of proteins, peptides, and other biomolecules when high purity is required.
  • Monodisperse 10µm porous beads yield high resolution, dynamic capacity, reproducibility and durability
  • Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q ion exchangers
  • Compatible with Ä;KTA design, FPLC System and other high-performance LC systems
  • MonoBeads™ are also available as Mono P for chromatofocusing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Mono Q HR 16/10
Mono Q HR 16/10 is a strong anion exchanger prepacked with MonoBeads. The column is an excellent choice for the polishing step in purification of proteins, peptides, and other biomolecules when high purity is required. The 20 mL bed volume of the column o
Monodisperse porous polystyrene/divinyl benzene beads
10μm, d50v is the median particle size of the cumulative volume distribution.
20 mL
16 mm
< 10 mL/min.
4 to 30°C, 20% Ethanol