Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Mono S PC, Columns Prepacked with MonoBeads™ Available on GSA/VA Contract for Federal Government customers only.

Use Mono Q or S precision columns for fast, sensitive, and high-resolution ion exchange separations of biomolecules at micropreparative scale. GE Healthcare Life Sciences™ Mono Q or S PC, Columns Prepacked with MonoBeads™ achieve maximum resolution from the efficiency of small, perfectly spherical monodispersed particles.

Manufacturer:  GE Healthcare 17516901

 View more versions of this product

Catalog No. 45-000-304


Add to cart

Description

Description

  • Maximum resolution achieved from efficiency of small spherical particles, optimally packed
  • Particles are coupled to Q or S ion exchangers with excellent sensitivities
  • Compatible with high-performance LC systems
  • Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid

  • Tricorn columns are optimized for the polishing step when high purity is required
  • PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
  • Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
  • Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
0.39 in.
Mono S ion exchange column
Small-scale polishing in purification of Proteins, Peptides and other biomolecules when high purity is required
100mm
Borosilicate Glass
40 bar
10μm
4° to 30°C
8mL
10mm
Each
4 in.
2 to 6mL/min.
O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-SO3
2 to 12
Monodisperse porous Polystyrene/Divinyl Benzene bead
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit