Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) Mono S PC, Columns Prepacked with MonoBeads™ Available on GSA/VA Contract for Federal Government customers only.

Use Mono Q or S precision columns for fast, sensitive, and high-resolution ion exchange separations of biomolecules at micropreparative scale. Cytiva Life Sciences™ Mono Q or S PC, Columns Prepacked with MonoBeads™ achieve maximum resolution from the efficiency of small, perfectly spherical monodispersed particles.

$2051.00 - $7923.00

Specifications

Buffer All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
pH Range 2 to 12
Storage Requirements 4° to 30°C
Particle Size 10μm
Support Type Monodisperse porous Polystyrene/Divinyl Benzene bead
View More Specs

Products 4
Catalog Number Mfr. No. Height (Metric) Max. Pressure Material Price Quantity & Availability  
Catalog Number Mfr. No. Height (Metric) Max. Pressure Material Price Quantity & Availability  
45-000-304 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
17516901
100mm 40 bar Borosilicate Glass
Each for $7,923.00
Add to cart
 
45-000-303 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
17516801
50mm 40 bar Borosilicate Glass
Each for $2,051.00
Add to cart
 
45-000-310
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
17518001
100mm 40 bar PEEK
Each for $2,252.00
Add to cart
 
45-000-124
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
17067201
50mm 50 bar Glass This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Maximum resolution achieved from efficiency of small spherical particles, optimally packed
  • Particles are coupled to Q or S ion exchangers with excellent sensitivities
  • Compatible with high-performance LC systems
  • Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid

  • Tricorn columns are optimized for the polishing step when high purity is required
  • PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
  • Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
  • Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
4° to 30°C
Monodisperse porous Polystyrene/Divinyl Benzene bead
2 to 12
10μm
O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-SO3
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit