Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAOA Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550998

Catalog No. PA550998


Add to cart

Description

Description

monoamine oxidase A Polyclonal Antibody detects endogenous levels of monoamine oxidase A protein.

This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome.
Specifications

Specifications

MAOA
Polyclonal
Unconjugated
MAOA
Liquid
MAOA
Synthesized peptide derived from the Internal region of human MAO-A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P21397, Q64133, P21396
MAOA, Amine oxidase [flavin-containing] A, Monoamine oxidase type A, MAO-A
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
17161, 29253, 4128
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit