Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monobenzone, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
103-16-2
C13H12O2
200.24
VYQNWZOUAUKGHI-UHFFFAOYSA-N
4-(benzyloxy)phenol
OC1=CC=C(OCC2=CC=CC=C2)C=C1
103-16-2
C13H12O2
200.24
VYQNWZOUAUKGHI-UHFFFAOYSA-N
4-(benzyloxy)phenol
OC1=CC=C(OCC2=CC=CC=C2)C=C1

M1506, 103-16-2, C13H12O2

Spectrum™ Monobenzone, USP is a depigmenting agent. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Ignition Residue 0.005
Loss on Drying 0.01
Melting Range 117-120°C
Quantity 25 kg
CAS Max % 1
Percent Purity 98 to 102%
Grade USP