Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner MONOBROMABIMANE 25 mg  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner F0030250MG
Monobromobimane reacts with thiols, sulfide, thiosulfate, and sulfite to generate a fluorescent product with an excitation/ emission spectra of 394/ 490 nm.
Catalog No. 50-720-324