Learn More
Sigma Aldrich Monodispersed nanodiamond particles
Catalog No. 900185100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
90-018-5100ML 100 mL
1 options
Catalog No. 90-018-5100ML Supplier Sigma Aldrich Supplier No. 900185100ML
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Detonation nanodiamonds find applications in polymer composites for providing improved mechanical properties, UV protection, and improved resistance to radiation. They also find biological applications extending from drug delivery to positron emission tomography and other imaging techniques.Do not freeze the materials.
Specifications
Quantity 100 mL
Synonym Detonation Nanodiamond; Nanodiamond