Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoethanolamine, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

141-43-5
C2H7NO
61.084
HZAXFHJVJLSVMW-UHFFFAOYSA-N
2-aminoethanol
700
CHEBI:16000

High quality chemicals for laboratory and specialized industrial use

CAS 141-43-5
Molecular Formula C2H7NO
Packaging Narrow Mouth Amber Glass Bottle
Synonym 2-aminoethanol
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
SMILES C(CO)N
IUPAC Name 2-aminoethanol
Molecular Weight (g/mol) 61.084
PubChem CID 700
ChEBI CHEBI:16000
Formula Weight 61.08
Specific Gravity 1L = 1.02kg
Flash Point 86.00°C
Quantity 500mL
Chemical Name or Material Monoethanolamine
Grade Baker Analyzed™ ACS Reagent
Show More Show Less