Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monopolin Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500131UV
Description
Monopolin Polyclonal antibody specifically detects Monopolin in Yeast samples. It is validated for Western BlotSpecifications
Monopolin | |
Polyclonal | |
50mM Sodium Borate | |
Synthetic peptide corresponding to a portion of human Monopolin. | |
0.1 mL | |
Cell Cycle and Replication | |
Yeast | |
Purified |
Western Blot | |
DyLight 350 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction