Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Montmorillonite, naturally occurring mineral, Thermo Scientific™

Catalog Number aa4253114
Click to view available options
Quantity:
25 g
1318-93-0
Al2H2O12Si4
360.307
MFCD00132796
GUJOJGAPFQRJSV-UHFFFAOYSA-N
montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
71586775
dialuminum;dioxosilane;oxygen(2-);hydrate
O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]
1318-93-0
Al2H2O12Si4
360.307
MFCD00132796
GUJOJGAPFQRJSV-UHFFFAOYSA-N
montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
71586775
dialuminum;dioxosilane;oxygen(2-);hydrate
O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]

Used in industrial chromatographic techniques, widely used in petroleum industry, catalyst carrier. It acts as a solid Lewis acid catalyst.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Montmorillonite
EINECSNumber 215-288-5
RTECSNumber QC4539200
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit