Learn More
Moore Medical APP OneTouch™ Ultra™ Test Strips
Quantitatively measures glucose in whole blood
Supplier:  Moore Medical 91402
Catalog No. 19-898-968
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • For use with OneTouch™ Ultra™ meter
  • 1μ blood sample
  • Easy-open flip top vial
  • Results in 5 seconds
  • Test on forearm or palm
Specifications
Specifications
25 Pack
5 sec.
1 μL