Learn More
Moore Medical Surgilast™ Dressing Retainer
Tubular elastic stretch net designed to serve as a secondary dressing
Manufacturer:  Moore Medical Corp 09803
 View more versions of this product
Catalog No. 19-898-036
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
25 yd.
Dressing Retainer
Elastic
22.86m
Ideal for securing burn or post-op dressings, incontinence pads and ducubitus ulcer dressings, easy to customize, allows maximum air-flow, for large hand, arm, leg, foot, self closing, serve as secondary dressing
Tubular