Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MooreBrand™ Adhesive Bandages

Made in numerous shapes, styles and sizes

Manufacturer:  Moore Medical Corp 68207

 View more versions of this product

Catalog No. 19-898-434


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

1.625 x 0.375 in. (4.1 x 0.95cm)
Yes
Butterfly
0.95cm
4.1cm
Adhesive bandage
4.1 × 0.95cm (1.625 × 0.375 in.)
For maximum protection
Yes
0.375 in.
1.625 in.
Plastic
100 Pack
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit