MooreBrand™ Berman Color-Coded Airway Kits and Replacements

Color-coded for fast and easy retrieval

Manufacturer: Moore Medical Corp 82133

 View more versions of this product

Catalog No. 19-898-737

  Check Availability
Add to Cart

    Specifications

    Color Lavender
    For Use With (Application) Triage, Emergency Response, First Aid
    Includes Each of all airways, Clear Plastic carrying case
    Size 100mm
    Product Type Airway Replacement

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit