Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical MooreBrand™ Emergency Blanket

Specifications

Color Yellow
For Use With (Application) Can be used as ground cover
View More Specs

Products 1
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19-898-441
View Documents
Moore Medical
68725
Poly with foam insulation This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Choice of aluminized mylar or poly with foam insulation
  • Can be used as ground cover

Specifications

Specifications

Yellow
Can be used as ground cover
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit