MooreBrand™ Emergency Blanket

Manufacturer: Moore Medical Corp 11687

 View all options for this product

Catalog No. 19-898-042


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Color Yellow
    Product Type Emergency blanket
    For Use With (Application) Can be used as ground cover
    Material Aluminized mylar

    • Choice of aluminized mylar or poly with foam insulation
    • Can be used as ground cover

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit