MooreBrand™ Table Paper

Gentle on patients yet durable to protect any type or size exam table from moisture, soil or other damage

Manufacturer: Moore Medical Corp 76992

 View more versions of this product

Catalog No. 19-898-574

  Check Availability
Add to Cart

    Specifications

    Color White
    Finish Crepe
    For Use With (Application) To protect any type or size exam table from moisture, soil or other damage
    Product Type Table Paper

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit