Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ MOPS, 0.2M buffer soln., pH 6.0, low endotoxin

Click to view available options
Quantity:
100mL
250mL
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
C1COCCN1CCCS(=O)(=O)O
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
CHEBI:44115
3-morpholin-4-ylpropane-1-sulfonic acid
C1COCCN1CCCS(=O)(=O)O
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 6.0
Color Colorless
Quantity 250mL
Chemical Name or Material MOPS, low endotoxin

TSCA : TSCA

storageNote1 : 2° to 8°C