Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma MOPS/EDTA Buffer, 10X Liquid Concentrate, Molecular Biology Grade, Calbiochem,
Prepared with DEPC for RNA applications
Manufacturer: MilliporeSigma 475916100ML
Specifications
MOPS/EDTA Buffer | |
10X concentrate contains 200mM MOPS, 50mM sodium acetate, 10mM EDTA | |
None Detected | |
None Detected | |
MOPS/EDTA Buffer |
C7H15NO4S | |
None Detected | |
Molecular Biology Grade | |
MOPS, Sodium Acetate, EDTA | |
100mL |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title