Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOPS sodium salt, 98%, ACROS Organics™
Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Glass bottle
Description
Biological buffer
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 98% |
Formula Weight | 231.25 |
Physical Form | Powder |
Percent Purity | 98% |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Color | White |
Quantity | 25g |
Show More |
Safety and Handling
TSCA : TSCA