Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
Packaging:
Poly Bottle

Chemical Identifiers

1132-61-2
C7H15NO4S
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
CHEBI:44115

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Molecular Biology Applications
CAS 1132-61-2
Molecular Formula C7H15NO4S
Packaging Poly Bottle
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
SMILES C1COCCN1CCCS(=O)(=O)O
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Molecular Weight (g/mol) 209.26
PubChem CID 70807
ChEBI CHEBI:44115
Formula Weight 209.26
Flash Point 116.00°C
Quantity 500g
Chemical Name or Material MOPS
Grade Ultrapure Bioreagent
Show More Show Less