Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS, Ultrapure Bioreagent, J.T. Baker™

Catalog No. p-5391236
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Molecular Biology Applications

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym mops, 4-morpholinepropanesulfonic acid, 3-morpholinopropanesulfonic acid, 3-n-morpholino propanesulfonic acid, 3-morpholinopropane-1-sulfonic acid, morpholinopropane sulfonic acid, morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 3-n-morpholinopropanesulfonic acid, unii-273bp63nv3
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Flash Point 116.00°C
Packaging Poly Bottle
Quantity 100 g
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
Formula Weight 209.26
Grade Ultrapure Bioreagent
Chemical Name or Material MOPS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.