Alfa Aesar™ MOPSO, 98%

Biological buffers, pH range 6.2 to 7.6

Manufacturer: Alfa Aesar™ 4413509

Quantity 10g

 View more versions of this product

Catalog No. AA4413509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 68399-77-9
    Molecular Formula C7H15NO5S
    Formula Weight 225.27
    MDL Number MFCD00009629
    Synonym beta-Hydroxy-4-morpholineproanesulfonic acid; 3-(4-Morpholino)-2-hydroxypropanesulfonic acid
    Chemical Name or Material MOPSO

    Specifications

    Formula Weight 225.27
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H15NO5S
    MDL Number MFCD00009629
    Synonym beta-Hydroxy-4-morpholineproanesulfonic acid; 3-(4-Morpholino)-2-hydroxypropanesulfonic acid
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 276° to 279°C
    TSCA TSCA
    CAS 68399-77-9
    Health Hazard 1 Warning
    Quantity 10g
    Chemical Name or Material MOPSO
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit