Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPSO, 98%

Catalog No. AA4413530
Click to view available options
Quantity:
50 g
250 g
1 kg

Biological buffers, pH range 6.2 to 7.6 | CAS: 68399-77-9 | C7H15NO5S | 225.259 g/mol

It is an important raw material used intermediate used in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals. Also used as biological buffers, pH range 6.2-7.6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68399-77-9
Molecular Formula C7H15NO5S
Molecular Weight (g/mol) 225.259
MDL Number MFCD00009629
InChI Key NUFBIAUZAMHTSP-UHFFFAOYSA-N
Synonym mopso, 2-hydroxy-3-morpholinopropanesulfonic acid, 2-hydroxy-3-morpholinopropane-1-sulfonic acid, 2-hydroxy-4-morpholinepropanesulphonic acid, 3-morpholino-2-hydroxypropanesulfonic acid, 3-n-morpholino-2-hydroxypropanesulfonic acid, 2-hydroxy-3-morpholin-4-yl propane-1-sulfonic acid, beta-hydroxy-4-morpholinepropanesulfonic acid, 3-4-morpholino-2-hydroxypropanesulfonic acid, 4-morpholinepropanesulfonic acid, .beta.-hydroxy
PubChem CID 109333
IUPAC Name 2-hydroxy-3-morpholin-4-ylpropane-1-sulfonic acid
SMILES C1COCCN1CC(CS(=O)(=O)O)O

Specifications

Melting Point 276°C to 279°C
Physical Form Powder
Quantity 250 g
Solubility Information Water solubility under desired conditions ca.112,6g/L at 20°C.
Formula Weight 225.27
Percent Purity 98%
Chemical Name or Material MOPSO
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 269-989-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.